Assay Details
Gene Symbol: Eif2b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133609.2 | NP_598293.2 | 11 - 12 | 1413 | 122 | |
XM_006238630.2 | XP_006238692.1 | 11 - 12 | 1384 | 122 | ||
GenBank mRNA | BC072507.1 | - | 11 - 12 | 1413 | 7099973 | 122 |
U38253.1 | - | 11 - 12 | 1393 | 122 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
