Assay Details
Gene Symbol: Mgst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134349.3 | NP_599176.1 | 2 - 3 | 195 | 68 | |
GenBank mRNA | BC063150.1 | - | 2 - 3 | 193 | 6922267 | 68 |
BP494884.1 | - | 2 - 3 | 194 | 68 | ||
CB711033.1 | - | 2 - 3 | 211 | 68 | ||
FJ179404.1 | - | 1 - 2 | 124 | 68 | ||
FQ209427.1 | - | 2 - 3 | 216 | 68 | ||
FQ209464.1 | - | 2 - 3 | 208 | 68 | ||
FQ209465.1 | - | 2 - 3 | 215 | 68 | ||
FQ211049.1 | - | 2 - 3 | 68 | |||
FQ212841.1 | - | 2 - 3 | 216 | 68 | ||
FQ218096.1 | - | 2 - 3 | 198 | 68 | ||
FQ218830.1 | - | 2 - 3 | 223 | 68 | ||
FQ219010.1 | - | 2 - 3 | 217 | 68 | ||
FQ219464.1 | - | 2 - 3 | 221 | 68 | ||
FQ219581.1 | - | 2 - 3 | 217 | 68 | ||
J03752.1 | - | 2 - 3 | 179 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
