Assay Details
Gene Symbol: Stambp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138531.2 | NP_612540.1 | 6 - 7 | 974 | 101 | |
XM_006236726.2 | XP_006236788.1 | 7 - 8 | 1641 | 101 | ||
XM_006236727.2 | XP_006236789.1 | 8 - 9 | 1759 | 101 | ||
XM_008763045.1 | XP_008761267.1 | 6 - 7 | 3302 | 101 | ||
XM_008763046.1 | XP_008761268.1 | 7 - 8 | 2322 | 101 | ||
XM_008763047.1 | XP_008761269.1 | 7 - 8 | 2739 | 101 | ||
XM_008763048.1 | XP_008761270.1 | 6 - 7 | 2621 | 101 | ||
GenBank mRNA | AY083159.1 | - | 5 - 6 | 798 | 101 | |
BC061711.1 | - | 6 - 7 | 974 | 5597943 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
