Assay Details
Gene Symbol: Ptgr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138863.2 | NP_620218.1 | 6 - 7 | 529 | 66 | |
GenBank mRNA | BC089775.1 | - | 6 - 7 | 529 | 66 | |
FQ218841.1 | - | 6 - 7 | 548 | 66 | ||
FQ221589.1 | - | 6 - 7 | 545 | 66 | ||
FQ222466.1 | - | 6 - 7 | 547 | 66 | ||
FQ229147.1 | - | 6 - 7 | 544 | 66 | ||
FQ229679.1 | - | 6 - 7 | 545 | 66 | ||
U66322.2 | - | 5 - 6 | 494 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
