Assay Details
Gene Symbol: Egfl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139104.1 | NP_620804.1 | 7 - 8 | 993 | 70 | |
XM_006233668.2 | XP_006233730.1 | 7 - 8 | 1096 | 70 | ||
XM_006233669.2 | XP_006233731.1 | 6 - 7 | 650 | 70 | ||
GenBank mRNA | AF223678.1 | - | 7 - 8 | 993 | 70 | |
BC078725.1 | - | 7 - 8 | 937 | 7127696 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
