Assay Details
Gene Symbol: Etfdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198742.2 | NP_942037.2 | 6 - 7 | 854 | 81 | |
XM_008761084.1 | XP_008759306.1 | 7 - 8 | 1080 | 81 | ||
GenBank mRNA | AY365022.1 | - | 6 - 7 | 680 | 81 | |
BC081890.1 | - | 6 - 7 | 854 | 81 | ||
FQ211230.1 | - | 6 - 7 | 911 | 81 | ||
FQ218772.1 | - | 7 - 8 | 1085 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
