Assay Details
Gene Symbol: Etfdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198742.2 | NP_942037.2 | 8 - 9 | 1148 | 85 | |
XM_006232507.1 | XP_006232569.1 | 7 - 8 | 1001 | 85 | ||
XM_008761084.1 | XP_008759306.1 | 9 - 10 | 1374 | 85 | ||
GenBank mRNA | AY365022.1 | - | 8 - 9 | 974 | 85 | |
BC081890.1 | - | 8 - 9 | 1148 | 85 | ||
FQ209791.1 | - | 2 - 3 | 178 | 85 | ||
FQ211230.1 | - | 8 - 9 | 1205 | 85 | ||
FQ218772.1 | - | 10 - 11 | 1515 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
