Assay Details
Gene Symbol: Esr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012754.1 | NP_036886.1 | 2 - 3 | 644 | 66 | |
XM_006240221.2 | XP_006240283.1 | 2 - 3 | 885 | 66 | ||
GenBank mRNA | AB012721.1 | - | 1 - 2 | 228 | 66 | |
AB190769.1 | - | 1 - 2 | 467 | 66 | ||
AB190770.1 | - | 1 - 2 | 467 | 66 | ||
AF042058.1 | - | 1 - 2 | 228 | 66 | ||
AF042059.1 | - | 1 - 2 | 228 | 66 | ||
AF042060.1 | - | 1 - 2 | 228 | 66 | ||
AF042061.1 | - | 1 - 2 | 228 | 66 | ||
U57439.1 | - | 2 - 3 | 644 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
