Assay Details
Gene Symbol: Esr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012754.1 | NP_036886.1 | 8 - 9 | 1685 | 72 | |
XM_006240221.2 | XP_006240283.1 | 9 - 10 | 1980 | 72 | ||
GenBank mRNA | AB012721.1 | - | 8 - 9 | 1323 | 72 | |
AB190769.1 | - | 7 - 8 | 1508 | 72 | ||
AB190770.1 | - | 8 - 9 | 1562 | 72 | ||
AF042058.1 | - | 7 - 8 | 1269 | 72 | ||
AF042059.1 | - | 8 - 9 | 1323 | 72 | ||
AF042060.1 | - | 6 - 7 | 1152 | 72 | ||
AF042061.1 | - | 7 - 8 | 1206 | 72 | ||
U57439.1 | - | 8 - 9 | 1685 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription by RNA polymerase II
ovarian follicle development
behavioral fear response
neuron migration
transcription, DNA-templated
regulation of transcription, DNA-templated
brain development
learning or memory
negative regulation of cell population proliferation
male gonad development
female gonad development
hormone-mediated apoptotic signaling pathway
response to xenobiotic stimulus
response to water deprivation
response to hormone
response to organic cyclic compound
response to activity
response to insecticide
cerebellum development
amygdala development
hypothalamus development
intracellular steroid hormone receptor signaling pathway
intracellular estrogen receptor signaling pathway
prostate gland development
response to nutrient levels
response to estradiol
response to testosterone
response to genistein
regulation of cell population proliferation
positive regulation of apoptotic process
steroid hormone mediated signaling pathway
regulation of neuron apoptotic process
response to estrogen
response to human chorionic gonadotropin
estrous cycle
response to ethanol
positive regulation of epidermal growth factor receptor signaling pathway
positive regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
negative regulation of behavior
negative regulation of smooth muscle cell proliferation
negative regulation of epithelial cell proliferation
positive regulation of DNA-binding transcription factor activity
Sertoli cell development
Sertoli cell proliferation
uterus development
vagina development
negative regulation of cell death
prostate gland epithelium morphogenesis
epithelial cell maturation involved in prostate gland development
negative regulation of androgen receptor signaling pathway
positive regulation of ERK1 and ERK2 cascade
cellular response to magnetism
cellular response to estradiol stimulus
cellular response to organic cyclic compound
cellular response to xenobiotic stimulus
response to dexamethasone
negative regulation of neuron death
response to salt
response to bisphenol A
negative regulation of feeding behavior
negative regulation of reactive oxygen species metabolic process
RNA polymerase II cis-regulatory region sequence-specific DNA binding
core promoter sequence-specific DNA binding
DNA binding
DNA-binding transcription factor activity
steroid hormone receptor activity
nuclear receptor activity
steroid binding
protein binding
zinc ion binding
enzyme binding
estrogen receptor activity
estrogen response element binding
hormone binding
peroxisome proliferator activated receptor binding
sequence-specific DNA binding
estradiol binding
steroid hormone binding
negative regulation of transcription by RNA polymerase II
ovarian follicle development
behavioral fear response
neuron migration
transcription, DNA-templated
regulation of transcription, DNA-templated
brain development
learning or memory
negative regulation of cell population proliferation
male gonad development
female gonad development
hormone-mediated apoptotic signaling pathway
response to xenobiotic stimulus
response to water deprivation
response to hormone
response to organic cyclic compound
response to activity
response to insecticide
cerebellum development
amygdala development
hypothalamus development
intracellular steroid hormone receptor signaling pathway
intracellular estrogen receptor signaling pathway
prostate gland development
response to nutrient levels
response to estradiol
response to testosterone
response to genistein
regulation of cell population proliferation
positive regulation of apoptotic process
steroid hormone mediated signaling pathway
regulation of neuron apoptotic process
response to estrogen
response to human chorionic gonadotropin
estrous cycle
response to ethanol
positive regulation of epidermal growth factor receptor signaling pathway
positive regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
negative regulation of behavior
negative regulation of smooth muscle cell proliferation
negative regulation of epithelial cell proliferation
positive regulation of DNA-binding transcription factor activity
Sertoli cell development
Sertoli cell proliferation
uterus development
vagina development
negative regulation of cell death
prostate gland epithelium morphogenesis
epithelial cell maturation involved in prostate gland development
negative regulation of androgen receptor signaling pathway
positive regulation of ERK1 and ERK2 cascade
cellular response to magnetism
cellular response to estradiol stimulus
cellular response to organic cyclic compound
cellular response to xenobiotic stimulus
response to dexamethasone
negative regulation of neuron death
response to salt
response to bisphenol A
negative regulation of feeding behavior
negative regulation of reactive oxygen species metabolic process
RNA polymerase II cis-regulatory region sequence-specific DNA binding
core promoter sequence-specific DNA binding
DNA binding
DNA-binding transcription factor activity
steroid hormone receptor activity
nuclear receptor activity
steroid binding
protein binding
zinc ion binding
enzyme binding
estrogen receptor activity
estrogen response element binding
hormone binding
peroxisome proliferator activated receptor binding
sequence-specific DNA binding
estradiol binding
steroid hormone binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P055961 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P049530 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371215 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8599 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566973 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899711 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245590 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401650 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683904 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477005 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830865 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351731 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108295 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690493 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752893 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586246 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560163 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858457 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528873 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875944 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371318 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578575 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69857 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321167 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702781 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882963 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214451 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124395 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177537 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92539 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103269 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123712 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P055962 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P055960 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P049530 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P055960 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P055962 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P049530 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P055960 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P055962 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P049530 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P442569 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 41882 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 83936 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 47322 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 47631 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 57184 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 77333 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10890658 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10890669 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10890659 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10890657 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10890664 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10890665 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10890661 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10890666 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10890667 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10890663 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10890656 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10890668 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10890670 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10890660 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10890654 | - | - |
Rat Genome 230 2.0 Array | 1369760_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1369760_a_at | - | - |
Affymetrix HT Rat Focus Array | 1369760_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1369760_a_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0600019084.rn.1 | TC0600002686.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600019085.rn.1 | TC0600002686.rn.1 | RTA TC |
Rat Expression Set 230 | 1369760_PM_a_at | - | - |
Rat Expression Set 230 | 1369760_a_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10890654 | - | - |
Rat Genome U34 Set | U57439_g_at | - | - |
Rat Genome U34 Set | U57439_at | - | - |
Rat Genome U34 Set | U57439_at | - | - |
Rat Genome U34 Set | U57439_g_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21927223 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1354998 | - | - |
RatRef-12 Expression BeadChip | ILMN_64780 | - | - |
