Assay Details
Gene Symbol: Acadvl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012891.2 | NP_037023.1 | 5 - 6 | 359 | 106 | |
GenBank mRNA | BC087036.1 | - | 5 - 6 | 380 | 7193535 | 106 |
D30647.1 | - | 5 - 6 | 361 | 106 | ||
FQ210754.1 | - | 5 - 6 | 408 | 106 | ||
FQ219329.1 | - | 5 - 6 | 404 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
