Assay Details
Gene Symbol: Lgals9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012977.1 | NP_037109.1 | 5 - 6 | 561 | 94 | |
GenBank mRNA | BC061566.1 | - | 5 - 6 | 522 | 94 | |
U59462.1 | - | 5 - 6 | 483 | 94 | ||
U67958.1 | - | 5 - 6 | 561 | 94 | ||
U72741.1 | - | 6 - 7 | 579 | 190 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
