Assay Details
Gene Symbol: Ahr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013149.2 | NP_037281.2 | 4 - 5 | 438 | 82 | |
GenBank mRNA | AF082124.1 | - | 4 - 5 | 467 | 82 | |
AF082125.1 | - | 4 - 5 | 467 | 82 | ||
AF082126.1 | - | 4 - 5 | 467 | 82 | ||
AJ821851.1 | - | 4 - 5 | 411 | 82 | ||
AM902281.1 | - | 4 - 5 | 438 | 82 | ||
AM902282.1 | - | 4 - 5 | 438 | 82 | ||
AM902283.1 | - | 4 - 5 | 438 | 82 | ||
AM902284.1 | - | 4 - 5 | 438 | 82 | ||
AM902285.1 | - | 4 - 5 | 438 | 82 | ||
AM902286.1 | - | 4 - 5 | 438 | 82 | ||
U09000.1 | - | 4 - 5 | 438 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
