Assay Details
Gene Symbol: Xylt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022296.2 | NP_071632.2 | 9 - 10 | 1936 | 110 | |
GenBank mRNA | AJ295749.2 | - | 9 - 10 | 1935 | 110 | |
BC105767.1 | - | 9 - 10 | 1936 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
