Assay Details
Gene Symbol: Mlst8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022404.2 | NP_071799.2 | 2 - 3 | 492 | 125 | |
GenBank mRNA | AF051155.2 | - | 2 - 3 | 491 | 125 | |
BC097319.1 | - | 2 - 3 | 170 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
