Assay Details
Gene Symbol: Oat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022521.3 | NP_071966.1 | 3 - 4 | 532 | 143 | |
GenBank mRNA | BC061551.1 | - | 3 - 4 | 535 | 143 | |
M11842.1 | - | 2 - 3 | 475 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
