Assay Details
Gene Symbol: Cd151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022523.1 | NP_071968.1 | 46 | 96 | ||
GenBank mRNA | AF192547.2 | - | 46 | 96 | ||
FQ222194.1 | - | 66 | 96 |
Gene Symbol: LOC100911730 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749017.3 | XP_003749065.1 | 1 - 2 | 106 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF192547.2, NM_022523.1, XM_003749017.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
