Assay Details
Gene Symbol: Ap2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031008.2 | NP_112270.2 | 19 - 20 | 2619 | 115 | |
XM_008760010.1 | XP_008758232.1 | 19 - 20 | 2674 | 115 | ||
GenBank mRNA | BC081786.1 | - | 19 - 20 | 2619 | 115 | |
X53773.1 | - | 2572 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
