Assay Details
Gene Symbol: Usf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031139.1 | NP_112401.1 | 2 - 3 | 618 | 131 | |
GenBank mRNA | AB035647.1 | - | 2 - 3 | 107 | 131 | |
AB035648.1 | - | 2 - 3 | 107 | 131 | ||
AB035649.1 | - | 2 - 3 | 107 | 131 | ||
AB047556.1 | - | 2 - 3 | 618 | 131 | ||
BC128735.1 | - | 2 - 3 | 112 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
