Assay Details
Gene Symbol: Mdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031151.2 | NP_112413.2 | 2 - 3 | 338 | 102 | |
GenBank mRNA | BC063165.1 | - | 2 - 3 | 338 | 6918633 | 102 |
DQ402951.1 | - | 1 - 2 | 102 | |||
FQ213494.1 | - | 2 - 3 | 291 | 102 | ||
FQ213817.1 | - | 2 - 3 | 255 | 102 | ||
FQ215385.1 | - | 2 - 3 | 298 | 102 | ||
FQ215647.1 | - | 2 - 3 | 299 | 102 | ||
FQ216782.1 | - | 2 - 3 | 329 | 102 | ||
FQ217232.1 | - | 2 - 3 | 290 | 102 | ||
FQ224847.1 | - | 2 - 3 | 235 | 102 | ||
FQ224979.1 | - | 2 - 3 | 317 | 102 | ||
X04240.1 | - | 2 - 3 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
