Assay Details
Gene Symbol: Mdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031151.2 | NP_112413.2 | 3 - 4 | 422 | 109 | |
GenBank mRNA | BC063165.1 | - | 3 - 4 | 422 | 6918633 | 109 |
DQ402951.1 | - | 2 - 3 | 256 | 109 | ||
FQ213494.1 | - | 3 - 4 | 375 | 109 | ||
FQ215385.1 | - | 3 - 4 | 382 | 109 | ||
FQ215647.1 | - | 3 - 4 | 383 | 109 | ||
FQ216782.1 | - | 3 - 4 | 413 | 109 | ||
FQ217232.1 | - | 3 - 4 | 374 | 109 | ||
FQ224847.1 | - | 3 - 4 | 319 | 109 | ||
FQ224979.1 | - | 3 - 4 | 401 | 109 | ||
X04240.1 | - | 3 - 4 | 363 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
