Assay Details
Gene Symbol: Slc7a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031341.1 | NP_112631.1 | 7 - 8 | 1301 | 67 | |
XM_006252020.2 | XP_006252082.1 | 7 - 8 | 1237 | 67 | ||
XM_006252021.2 | XP_006252083.1 | 10 - 11 | 1706 | 67 | ||
XM_006252023.2 | XP_006252085.1 | 7 - 8 | 1242 | 67 | ||
XM_006252024.2 | XP_006252086.1 | 7 - 8 | 1931 | 67 | ||
XM_008770705.1 | XP_008768927.1 | 9 - 10 | 1649 | 67 | ||
GenBank mRNA | AB020520.1 | - | 7 - 8 | 1301 | 67 | |
AF200684.1 | - | 6 - 7 | 1109 | 67 | ||
BC091142.1 | - | 7 - 8 | 1216 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
