Assay Details
Gene Symbol: Stk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031735.1 | NP_113923.1 | 3 - 4 | 316 | 140 | |
XM_008765458.1 | XP_008763680.1 | 3 - 4 | 494 | 140 | ||
XM_008765459.1 | XP_008763681.1 | 3 - 4 | 494 | 140 | ||
XM_008765460.1 | XP_008763682.1 | 3 - 4 | 183 | 140 | ||
XM_008765461.1 | XP_008763683.1 | 3 - 4 | 494 | 140 | ||
XM_008765462.1 | XP_008763684.1 | 3 - 4 | 494 | 140 | ||
GenBank mRNA | AJ001529.1 | - | 3 - 4 | 316 | 140 | |
BC161844.1 | - | 3 - 4 | 399 | 9027884 | 140 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008765460.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
