Assay Details
Gene Symbol: Ceacam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033860.1 | NP_001029032.1 | 8 - 9 | 1504 | 66 | |
NM_001033861.1 | NP_001029033.1 | 6 - 7 | 1324 | 66 | ||
NM_001033862.1 | NP_001029034.1 | 7 - 8 | 1451 | 66 | ||
NM_031755.2 | NP_113943.1 | 7 - 8 | 1377 | 66 | ||
GenBank mRNA | AJ277104.1 | - | 7 - 8 | 1339 | 66 | |
AJ277105.1 | - | 6 - 7 | 1286 | 66 | ||
BC061740.1 | - | 8 - 9 | 1490 | 66 | ||
J04963.1 | - | 8 - 9 | 1511 | 66 | ||
X91137.1 | - | 7 - 8 | 1413 | 66 | ||
Z12019.1 | - | 7 - 8 | 1414 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
