Assay Details
Gene Symbol: Hagh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033349.2 | NP_203500.2 | 6 - 7 | 676 | 101 | |
XM_006245899.1 | XP_006245961.1 | 6 - 7 | 653 | 101 | ||
XM_006245900.1 | XP_006245962.1 | 5 - 6 | 567 | 101 | ||
GenBank mRNA | BC097301.1 | - | 6 - 7 | 628 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
