Assay Details
Gene Symbol: Rab27b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053459.1 | NP_445911.1 | 3 - 4 | 470 | 62 | |
XM_006254930.2 | XP_006254992.1 | 4 - 5 | 432 | 62 | ||
GenBank mRNA | AF325693.1 | - | 3 - 4 | 470 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
