Assay Details
Gene Symbol: Dao | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053626.1 | NP_446078.1 | 6 | 603 | 68 | |
GenBank mRNA | AB003400.1 | - | 6 | 603 | 68 | |
BC088395.1 | - | 5 | 518 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
