Assay Details
Gene Symbol: Pitpnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053742.1 | NP_446194.1 | 1 - 2 | 47 | 72 | |
XM_006249522.2 | XP_006249584.1 | 1 - 2 | 91 | 72 | ||
XM_006249523.2 | XP_006249585.1 | 1 - 2 | 91 | 72 | ||
GenBank mRNA | BC061538.1 | - | 1 - 2 | 79 | 72 | |
D21132.1 | - | 1 - 2 | 47 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC061538.1, D21132.1, NM_053742.1, XM_006249522.2, XM_006249523.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
