Assay Details
Gene Symbol: Cfh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130409.2 | NP_569093.2 | 20 - 21 | 3372 | 170 | |
XM_008769531.1 | XP_008767753.1 | 19 - 20 | 3398 | 170 | ||
XM_008769532.1 | XP_008767754.1 | 11 - 12 | 1930 | 170 | ||
GenBank mRNA | AJ320522.1 | - | 3349 | 173 | ||
BC083174.1 | - | 1743 | 170 | |||
CF110979.1 | - | 4 - 5 | 728 | 170 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
