Assay Details
Gene Symbol: Cd79b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133533.2 | NP_598217.1 | 4 - 5 | 605 | 76 | |
GenBank mRNA | BC088115.1 | - | 4 - 5 | 605 | 76 | |
FQ226266.1 | - | 4 - 5 | 602 | 76 | ||
FQ227061.1 | - | 4 - 5 | 600 | 76 | ||
FQ230261.1 | - | 4 - 5 | 596 | 76 | ||
FQ232054.1 | - | 4 - 5 | 596 | 76 | ||
FQ232680.1 | - | 4 - 5 | 76 | |||
FQ232790.1 | - | 4 - 5 | 619 | 76 | ||
FQ233507.1 | - | 4 - 5 | 594 | 76 | ||
FQ234082.1 | - | 4 - 5 | 579 | 76 | ||
FQ234487.1 | - | 4 - 5 | 594 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
