Assay Details
Gene Symbol: Rph3al | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133591.2 | NP_598275.1 | 6 - 7 | 731 | 71 | |
XM_008767941.1 | XP_008766163.1 | 6 - 7 | 1092 | 71 | ||
XM_008767942.1 | XP_008766164.1 | 6 - 7 | 1157 | 71 | ||
XM_008767943.1 | XP_008766165.1 | 5 - 6 | 1005 | 71 | ||
GenBank mRNA | AF022774.1 | - | 5 - 6 | 657 | 71 | |
BC091126.1 | - | 6 - 7 | 731 | 7380477 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
