Assay Details
Gene Symbol: Kcnh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133610.2 | NP_598294.1 | 4 - 5 | 701 | 69 | |
GenBank mRNA | AF185637.1 | - | 4 - 5 | 660 | 69 | |
AJ250280.1 | - | 4 - 5 | 703 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
