Assay Details
Gene Symbol: Ctsd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134334.2 | NP_599161.2 | 4 - 5 | 584 | 93 | |
GenBank mRNA | BC062032.1 | - | 4 - 5 | 584 | 93 | |
X54467.1 | - | 4 - 5 | 529 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
