Assay Details
Gene Symbol: Gpr156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153295.1 | NP_695207.1 | 8 - 9 | 1082 | 80 | |
GenBank mRNA | AF488741.1 | - | 8 - 9 | 1082 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P156762 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449525 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103559 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682722 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606627 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851904 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702030 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30525 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539652 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115030 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53820 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634661 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721589 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738082 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61329 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728108 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376765 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249672 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360272 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251809 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508762 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353719 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95283 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14261 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777266 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130324 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465113 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226235 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121074 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10583 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34785 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P156762 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P156762 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P156762 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P339477 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 10931 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 5519 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 24498 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 86136 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10754205 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10754204 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10754206 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10754207 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10754203 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10754209 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10754210 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10754208 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10754202 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10754201 | - | - |
Affymetrix Rat Clariom S Assay | PSR1100010053.rn.1 | TC1100001659.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10754201 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20783495 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1368664 | - | - |
RatRef-12 Expression BeadChip | ILMN_66462 | - | - |
