Assay Details
Gene Symbol: Havcr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173149.2 | NP_775172.1 | 2 - 3 | 987 | 81 | |
XM_006246163.2 | XP_006246225.1 | 3 - 4 | 632 | 81 | ||
XM_008767665.1 | XP_008765887.1 | 4 - 5 | 684 | 81 | ||
XM_008767666.1 | XP_008765888.1 | 3 - 4 | 1269 | 81 | ||
GenBank mRNA | AF035963.1 | - | 2 - 3 | 988 | 81 | |
BC061820.1 | - | 3 - 4 | 427 | 5599247 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
