Assay Details
Gene Symbol: Havcr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173149.2 | NP_775172.1 | 4 - 5 | 1204 | 105 | |
XM_006246163.2 | XP_006246225.1 | 5 - 6 | 849 | 105 | ||
XM_008767665.1 | XP_008765887.1 | 6 - 7 | 901 | 105 | ||
XM_008767666.1 | XP_008765888.1 | 5 - 6 | 1486 | 105 | ||
GenBank mRNA | AF035963.1 | - | 4 - 5 | 1205 | 105 | |
BC061820.1 | - | 5 - 6 | 644 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
