Assay Details
Gene Symbol: Havcr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173149.2 | NP_775172.1 | 6 - 7 | 1371 | 98 | |
XM_006246163.2 | XP_006246225.1 | 7 - 8 | 1016 | 98 | ||
XM_008767665.1 | XP_008765887.1 | 8 - 9 | 1068 | 98 | ||
XM_008767666.1 | XP_008765888.1 | 7 - 8 | 1653 | 98 | ||
GenBank mRNA | AF035963.1 | - | 6 - 7 | 1372 | 98 | |
BC061820.1 | - | 7 - 8 | 811 | 5599247 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
