Assay Details
Gene Symbol: Chst15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173310.3 | NP_775432.3 | 4 - 5 | 1206 | 92 | |
XM_006230430.2 | XP_006230492.1 | 5 - 6 | 2773 | 92 | ||
XM_006230431.2 | XP_006230493.1 | 5 - 6 | 3513 | 92 | ||
XM_006230433.2 | XP_006230495.1 | 5 - 6 | 3008 | 92 | ||
XM_008759868.1 | XP_008758090.1 | 5 - 6 | 1769 | 92 | ||
GenBank mRNA | AB086953.1 | - | 4 - 5 | 1206 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
