Assay Details
Gene Symbol: Adamts5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198761.1 | NP_942056.1 | 7 - 8 | 2223 | 115 | |
GenBank mRNA | AY382879.1 | - | 7 - 8 | 2223 | 115 | |
FQ228283.1 | - | 2616 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P508162 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942052 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945905 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716910 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862930 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618096 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455770 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483444 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108909 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736392 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324758 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935082 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641107 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637118 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436307 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106690 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90271 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618662 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113771 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142399 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61243 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151450 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P508162 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P508162 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P508162 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P508162 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 55683 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 39653 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 27687 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 38837 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10752857 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10752856 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10752854 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10752853 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10752860 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10752855 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10752859 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10752858 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10752852 | - | - |
Rat Genome 230 2.0 Array | 1394483_PM_at | - | - |
Rat Genome 230 2.0 Array | 1394483_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1100007041.rn.1 | TC1100001212.rn.1 | RTA TC |
Rat Expression Set 230 | 1394483_PM_at | - | - |
Rat Expression Set 230 | 1394483_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10752852 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21182826 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1366387 | - | - |
RatRef-12 Expression BeadChip | ILMN_61299 | - | - |
