Assay Details
Gene Symbol: Spint2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199087.1 | NP_954518.1 | 1 - 2 | 388 | 56 | |
GenBank mRNA | BC061768.1 | - | 1 - 2 | 388 | 5621353 | 56 |
FQ229514.1 | - | 2 - 3 | 287 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
