Assay Details
Gene Symbol: Tor1aip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199100.2 | NP_954531.1 | 2 - 3 | 279 | 77 | |
XM_006250043.2 | XP_006250105.1 | 1 - 2 | 184 | 77 | ||
XM_006250044.2 | XP_006250106.1 | 1 - 2 | 182 | 77 | ||
XM_006250045.2 | XP_006250107.1 | 1 - 2 | 182 | 77 | ||
GenBank mRNA | BC061831.1 | - | 2 - 3 | 236 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
