Assay Details
Gene Symbol: Tor1aip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199100.2 | NP_954531.1 | 4 - 5 | 1182 | 77 | |
XM_006250043.2 | XP_006250105.1 | 3 - 4 | 1087 | 77 | ||
XM_006250044.2 | XP_006250106.1 | 3 - 4 | 1085 | 77 | ||
XM_006250045.2 | XP_006250107.1 | 3 - 4 | 1085 | 77 | ||
GenBank mRNA | BC061831.1 | - | 4 - 5 | 1139 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P160673 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833030 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78794 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208758 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106262 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310218 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440897 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687318 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956269 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63070 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675551 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288028 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182667 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 83061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46379 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P160673 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P160673 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P160673 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P163176 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 35836 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 81109 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 23678 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10764828 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764834 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764832 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764831 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764830 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764836 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764829 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764835 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764833 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10764827 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10764837 | - | - |
Rat Genome 230 2.0 Array | 1375969_PM_at | - | - |
Rat Genome 230 2.0 Array | 1374798_at | - | - |
Rat Genome 230 2.0 Array | 1375969_at | - | - |
Rat Genome 230 2.0 Array | 1383368_PM_at | - | - |
Rat Genome 230 2.0 Array | 1381980_PM_at | - | - |
Rat Genome 230 2.0 Array | 1381980_at | - | - |
Rat Genome 230 2.0 Array | 1383368_at | - | - |
Rat Genome 230 2.0 Array | 1374798_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1375969_at | - | - |
Affymetrix HT Rat Focus Array | 1374798_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1381980_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1375969_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1383368_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1374798_at | - | - |
Affymetrix HT Rat Focus Array | 1381980_at | - | - |
Affymetrix HT Rat Focus Array | 1383368_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1300003724.rn.1 | TC1300000541.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300003721.rn.1 | TC1300000541.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300003720.rn.1 | TC1300000541.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300003723.rn.1 | TC1300000541.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300003719.rn.1 | TC1300000541.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300003722.rn.1 | TC1300000541.rn.1 | RTA TC |
Rat Expression Set 230 | 1374798_PM_at | - | - |
Rat Expression Set 230 | 1374798_at | - | - |
Rat Expression Set 230 | 1375969_at | - | - |
Rat Expression Set 230 | 1375969_PM_at | - | - |
Rat Expression Set 230 | 1381980_PM_at | - | - |
Rat Expression Set 230 | 1381980_at | - | - |
Rat Expression Set 230 | 1383368_at | - | - |
Rat Expression Set 230 | 1383368_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764827 | - | - |
Rat Genome U34 Set | rc_AA892284_at | - | - |
Rat Genome U34 Set | rc_AA997589_at | - | - |
Rat Genome U34 Set | rc_AA924151_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20722858 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1362870 | - | - |
RatRef-12 Expression BeadChip | ILMN_66569 | - | - |
