Assay Details
Gene Symbol: Dld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199385.2 | NP_955417.1 | 7 - 8 | 658 | 72 | |
XM_008764590.1 | XP_008762812.1 | 7 - 8 | 675 | 72 | ||
GenBank mRNA | BC062069.1 | - | 7 - 8 | 662 | 72 | |
FM093110.1 | - | 2 - 3 | 236 | 72 | ||
FQ215299.1 | - | 7 - 8 | 665 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
