Assay Details
Gene Symbol: Mylpf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012605.2 | NP_036737.1 | 2 - 3 | 131 | 70 | |
GenBank mRNA | BI278596.1 | - | 5 - 6 | 70 | ||
CB770854.1 | - | 2 - 3 | 141 | 70 | ||
FQ215085.1 | - | 2 - 3 | 164 | 70 | ||
FQ216891.1 | - | 2 - 3 | 147 | 70 | ||
FQ216934.1 | - | 2 - 3 | 147 | 70 | ||
FQ217098.1 | - | 2 - 3 | 147 | 70 | ||
FQ217155.1 | - | 2 - 3 | 147 | 70 | ||
FQ217219.1 | - | 2 - 3 | 147 | 70 | ||
FQ217259.1 | - | 2 - 3 | 147 | 70 | ||
FQ217349.1 | - | 1 - 2 | 71 | 70 | ||
FQ217365.1 | - | 2 - 3 | 146 | 70 | ||
FQ217368.1 | - | 2 - 3 | 163 | 70 | ||
FQ217533.1 | - | 2 - 3 | 175 | 70 | ||
FQ217703.1 | - | 2 - 3 | 147 | 70 | ||
FQ217944.1 | - | 2 - 3 | 147 | 70 | ||
FQ218017.1 | - | 2 - 3 | 147 | 70 | ||
FQ223762.1 | - | 2 - 3 | 128 | 70 | ||
FQ223868.1 | - | 2 - 3 | 147 | 70 | ||
FQ223911.1 | - | 2 - 3 | 163 | 70 | ||
FQ223966.1 | - | 2 - 3 | 147 | 70 | ||
FQ223985.1 | - | 2 - 3 | 147 | 70 | ||
FQ224231.1 | - | 2 - 3 | 147 | 70 | ||
FQ224391.1 | - | 2 - 3 | 145 | 70 | ||
FQ224404.1 | - | 2 - 3 | 145 | 70 | ||
FQ224511.1 | - | 2 - 3 | 147 | 70 | ||
FQ224720.1 | - | 2 - 3 | 147 | 70 | ||
FQ224781.1 | - | 2 - 3 | 163 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
