Assay Details
Gene Symbol: Map6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017204.1 | NP_058900.1 | 2 | 1745 | 61 | |
XM_006229755.2 | XP_006229817.1 | 1 | 2075 | 61 | ||
GenBank mRNA | AJ002556.1 | - | 1 | 1764 | 61 | |
BC078848.1 | - | 1 | 1623 | 61 | ||
X93495.1 | - | 2 | 1745 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
