Assay Details
Gene Symbol: Parva | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020656.2 | NP_065707.2 | 7 - 8 | 718 | 85 | |
GenBank mRNA | AF264765.2 | - | 7 - 8 | 718 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
