Assay Details
Gene Symbol: Arl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022385.3 | NP_071780.1 | 3 - 4 | 308 | 131 | |
GenBank mRNA | BC061553.1 | - | 391 | 131 | ||
FQ226660.1 | - | 3 - 4 | 364 | 131 | ||
U12402.1 | - | 3 - 4 | 240 | 131 | ||
X76920.1 | - | 3 - 4 | 308 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
