Assay Details
Gene Symbol: Il18bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053374.2 | NP_445826.2 | 2 - 3 | 230 | 80 | |
XM_008759805.1 | XP_008758027.1 | 2 - 3 | 2388 | 80 | ||
GenBank mRNA | AF154569.1 | - | 1 - 2 | 41 | 80 | |
CB809932.1 | - | 2 - 3 | 241 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF154569.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
