Assay Details
Gene Symbol: Cdc5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053527.1 | NP_445979.1 | 12 - 13 | 1755 | 103 | |
GenBank mRNA | AF000578.2 | - | 12 - 13 | 1755 | 103 | |
BC087007.1 | - | 13 - 14 | 1802 | 103 | ||
BC161839.1 | - | 12 - 13 | 1719 | 9101355 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
