Assay Details
Gene Symbol: Folr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133527.1 | NP_598211.1 | 4 - 5 | 514 | 93 | |
XM_006229842.2 | XP_006229904.1 | 4 - 5 | 635 | 93 | ||
GenBank mRNA | AF219904.1 | - | 4 - 5 | 514 | 93 | |
BC157811.1 | - | 3 - 4 | 497 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
