Assay Details
Gene Symbol: Mapkapk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012127.1 | NP_001012127.1 | 5 - 6 | 794 | 80 | |
XM_006243776.2 | XP_006243838.1 | 5 - 6 | 884 | 80 | ||
XM_008766531.1 | XP_008764753.1 | 5 - 6 | 893 | 80 | ||
GenBank mRNA | BC081974.1 | - | 5 - 6 | 794 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
